siRNA Details
Gene Symbol: Sgcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011891.4 | NP_036021.1 | 3, 4 | 349 | ||
GenBank mRNA | AB024923.1 | - | 3, 4 | 570 | ||
AK040974.1 | - | 3, 4 | 351 | |||
BC130019.1 | - | 3, 4 | 339 | 40140624 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
