siRNA Details
| Gene Symbol: Slc35a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011895.3 | NP_036025.2 | 2 | 321 | ||
| GenBank mRNA | AK144170.1 | - | 2 | 315 | ||
| BC012252.1 | - | 2 | 236 | 4190458 | ||
| CF172178.1 | - | 1 | 127 | |||
| Z71268.1 | - | 2 | 142 | |||

Slc35a1
Genomic Map