siRNA Details
| Gene Symbol: Spry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF176903.1 | - | 3 | 1821 | ||
| AK014578.1 | - | 3 | 1839 | |||
| AK140405.1 | - | 3 | 1833 | |||
| BC039139.1 | - | 4 | 1679 | 3672353 | ||
| BC051977.1 | - | 2 | 2125 | |||
| BC053428.1 | - | 3 | 1823 | 6506766 | ||
| BC069914.1 | - | 1 | 1342 | 5251876 | ||

Spry1
Genomic Map