siRNA Details
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112705.2 | NP_001106176.1 | 4 | 734 | ||
| NM_001294329.1 | NP_001281258.1 | 4 | 734 | |||
| NM_001294330.1 | NP_001281259.1 | 4 | 734 | |||
| NM_001294331.1 | NP_001281260.1 | 5 | 843 | |||
| NM_001294334.1 | NP_001281263.1 | 4 | 734 | |||
| GenBank mRNA | AF045252.1 | - | 5 | 849 | ||
| AF045253.1 | - | 3 | 181 | |||
| AK014829.1 | - | 4 | 372 | |||
| AK048492.1 | - | 1 | 1234 | |||
| AK089925.1 | - | 4 | 262 | |||
| AK165667.1 | - | 4 | 737 | |||
| BC066198.1 | - | 4 | 279 | |||

Tlk2
Genomic Map