siRNA Details
| Gene Symbol: Ubl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011908.2 | NP_036038.1 | 3 | 975 | ||
| GenBank mRNA | AF044222.1 | - | 3 | 917 | ||
| AF044223.1 | - | 3 | 236 | |||
| AK134598.1 | - | 3 | 343 | |||
| AK145952.1 | - | 3 | 654 | |||
| BC024507.1 | - | 5 | 825 | 4983604 | ||
| BC025595.1 | - | 3 | 630 | 5342735 | ||
| BC043729.1 | - | 5 | 825 | 4983604 | ||

Ubl3
Genomic Map