siRNA Details
| Gene Symbol: Nelfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011914.2 | NP_036044.1 | 6 | 814 | ||
| GenBank mRNA | AF101435.1 | - | 6 | 772 | ||
| AK019191.1 | - | 6 | 880 | |||
| AK078760.1 | - | 6 | 814 | |||
| AK088384.1 | - | 6 | 830 | |||
| AK144173.1 | - | 828 | ||||
| AK146185.1 | - | 6 | 866 | |||
| AK180606.1 | - | 2 | 80 | |||
| BC038003.1 | - | 6 | 815 | 4912719 | ||

Nelfa
Genomic Map