siRNA Details
| Gene Symbol: Nelfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011914.2 | NP_036044.1 | 11 | 2142 | ||
| GenBank mRNA | AF101435.1 | - | 11 | 2089 | ||
| AK012673.1 | - | 1 | 711 | |||
| AK019191.1 | - | 11 | 2208 | |||
| AK088384.1 | - | 11 | 2157 | |||
| AK144173.1 | - | 2154 | ||||
| AK146185.1 | - | 11 | 2192 | |||
| BC018423.1 | - | 2 | 757 | |||
| BC038003.1 | - | 11 | 2143 | 4912719 | ||

Nelfa
Genomic Map