siRNA Details
| Gene Symbol: Ing1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF177755.1 | - | 2 | 1116 | ||
| AF177756.1 | - | 2 | 1101 | |||
| AF177757.1 | - | 2 | 1974 | |||
| AK035490.1 | - | 2 | 1131 | |||
| AK076482.1 | - | 2 | 1024 | |||
| AK141466.1 | - | 2 | 1024 | |||
| AK145553.1 | - | 2 | 1026 | |||
| AK157424.1 | - | 2 | 1123 | |||
| BC016573.1 | - | 2 | 1516 | |||

Ing1
Genomic Map