siRNA Details
| Gene Symbol: Aldh1a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011921.2 | NP_036051.1 | 13 | 1707 | ||
| GenBank mRNA | BC032880.1 | - | 13 | 1684 | ||
| BC046315.1 | - | 13 | 1681 | 6314273 | ||
| BC139412.1 | - | 13 | 1684 | 8862434 | ||
| BC139413.1 | - | 13 | 1684 | 8862435 | ||
| U96401.1 | - | 13 | 1707 | |||

Aldh1a7
Genomic Map