siRNA Details
| Gene Symbol: Adgre5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163030.1 | NP_001156502.1 | 5 | 569 | ||
| NM_011925.2 | NP_036055.2 | 6 | 704 | |||
| GenBank mRNA | CA319344.1 | - | 4 | 445 | ||
| CD553351.1 | - | 5 | 422 | |||
| Y18365.1 | - | 6 | 562 | |||
More Information
Set Membership: |
|

Adgre5
Genomic Map