siRNA Details
| Gene Symbol: Clcn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011929.2 | NP_036059.1 | 16 | 1556 | ||
| GenBank mRNA | AK145053.1 | - | 16 | 1601 | ||
| AK165362.1 | - | 16 | 1606 | |||
| AK220319.1 | - | 10 | 1046 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clcn6| Gene Symbol: Clcn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011929.2 | NP_036059.1 | 16 | 1556 | ||
| GenBank mRNA | AK145053.1 | - | 16 | 1601 | ||
| AK165362.1 | - | 16 | 1606 | |||
| AK220319.1 | - | 10 | 1046 | |||