siRNA Details
| Gene Symbol: Dapp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011932.2 | NP_036062.1 | 4 | 516 | ||
| GenBank mRNA | AF163255.1 | - | 4 | 470 | ||
| AF186023.1 | - | 4 | 491 | |||
| AK076886.1 | - | 2 | 69 | |||
| AK145745.1 | - | 4 | 482 | |||
| AK146180.1 | - | 4 | 485 | |||
| AK169584.1 | - | 3 | 381 | |||
| BC014759.1 | - | 4 | 512 | 4486834 | ||

Dapp1
Genomic Map