siRNA Details
| Gene Symbol: Esrrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159500.1 | NP_001152972.1 | 7 | 4093 | ||
| NM_011934.4 | NP_036064.3 | 7 | 3992 | |||
| GenBank mRNA | AK032025.1 | - | 7 | 3800 | ||
| AK052256.1 | - | 7 | 4096 | |||
| BC044858.1 | - | 6 | 3662 | |||

Esrrb
Genomic Map