siRNA Details
| Gene Symbol: Map3k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011947.3 | NP_036077.1 | 3, 4 | 202 | ||
| GenBank mRNA | AK137127.1 | - | 3, 4 | 488 | ||
| AK220450.1 | - | 3, 4 | 496 | |||
| BC023781.1 | - | 3, 4 | 206 | 5346982 | ||
| U43187.1 | - | 3, 4 | 505 | |||

Map3k3
Genomic Map