siRNA Details
| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011958.3 | NP_036088.3 | 14 | 1741 | ||
| GenBank mRNA | AK076324.1 | - | 14 | 1742 | ||
| AK084326.1 | - | 14 | 1710 | |||
| AK169291.1 | - | 14 | 1725 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Orc4| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011958.3 | NP_036088.3 | 14 | 1741 | ||
| GenBank mRNA | AK076324.1 | - | 14 | 1742 | ||
| AK084326.1 | - | 14 | 1710 | |||
| AK169291.1 | - | 14 | 1725 | |||