siRNA Details
| Gene Symbol: Psma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011967.3 | NP_036097.1 | 8, 9 | 997 | ||
| GenBank mRNA | AF019661.1 | - | 8, 9 | 712 | ||
| AK143468.1 | - | 8, 9 | 999 | |||
| AK160530.1 | - | 8, 9 | 757 | |||
| AK160550.1 | - | 8, 9 | 751 | |||
| AK160741.1 | - | 8, 9 | 760 | |||
| AK161680.1 | - | 8, 9 | 745 | |||
| BC007181.1 | - | 8, 9 | 744 | |||
| BC010709.1 | - | 8, 9 | 733 | 2648635 | ||
| BC083342.1 | - | 8, 9 | 730 | 5700963 | ||
| BC108368.1 | - | 8, 9 | 727 | 3596530 | ||

Psma5
Genomic Map