siRNA Details
| Gene Symbol: Vnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011979.2 | NP_036109.3 | 6 | 1431 | ||
| GenBank mRNA | AJ132103.1 | - | 6 | 1421 | ||
| BC111521.2 | - | 6 | 1403 | 40047284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vnn3| Gene Symbol: Vnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011979.2 | NP_036109.3 | 6 | 1431 | ||
| GenBank mRNA | AJ132103.1 | - | 6 | 1421 | ||
| BC111521.2 | - | 6 | 1403 | 40047284 | ||