siRNA Details
| Gene Symbol: Zfp146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ224763.1 | - | 2 | 752 | ||
| BC127039.1 | - | 1 | 231 | 40051941 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp146| Gene Symbol: Zfp146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ224763.1 | - | 2 | 752 | ||
| BC127039.1 | - | 1 | 231 | 40051941 | ||