siRNA Details
| Gene Symbol: Homer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146153.1 | NP_001139625.1 | 3 | 277 | ||
| NM_011984.2 | NP_036114.2 | 3 | 277 | |||
| GenBank mRNA | AF093261.1 | - | 1 | 104 | ||
| AK155463.1 | - | 1 | 2456 | |||
| BC005773.1 | - | 3 | 226 | 3602414 | ||
| BC095961.1 | - | 3 | 232 | |||
| CX208892.1 | - | 3 | 256 | |||

Homer3
Genomic Map