siRNA Details
| Gene Symbol: Dpysl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011993.4 | NP_036123.3 | 2, 3 | 247 | ||
| GenBank mRNA | AB006715.1 | - | 2, 3 | 176 | ||
| AK090197.1 | - | 2, 3 | 221 | |||
| AK145638.1 | - | 1, 2 | 350 | |||
| AK165694.1 | - | 2, 3 | 248 | |||
| BC046333.1 | - | 2, 3 | 221 | |||
| BC090674.1 | - | 220 | ||||
| BC138573.1 | - | 2, 3 | 165 | 8861593 | ||
| BC138574.1 | - | 2, 3 | 165 | 8861594 | ||
| Y09079.1 | - | 2, 3 | 226 | |||

Dpysl4
Genomic Map