siRNA Details
| Gene Symbol: Dpysl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011993.4 | NP_036123.3 | 14 | 1900 | ||
| GenBank mRNA | AB006715.1 | - | 14 | 1829 | ||
| AK090197.1 | - | 13 | 2428 | |||
| AK145638.1 | - | 13 | 2003 | |||
| AK165694.1 | - | 14 | 1901 | |||
| BC046333.1 | - | 13 | 3453 | |||
| BC090674.1 | - | 1872 | ||||
| BC138573.1 | - | 14 | 1818 | 8861593 | ||
| BC138574.1 | - | 14 | 1818 | 8861594 | ||
| Y09079.1 | - | 14 | 1879 | |||

Dpysl4
Genomic Map