siRNA Details
| Gene Symbol: Clec4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170332.1 | NP_001163803.1 | 7 | 2028 | ||
| NM_001170333.1 | NP_001163804.1 | 7 | 2180 | |||
| NM_011999.4 | NP_036129.1 | 6 | 2108 | |||
| GenBank mRNA | AK049002.1 | - | 6 | 2031 | ||

Clec4a2
Genomic Map