siRNA Details
| Gene Symbol: Cops7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012003.2 | NP_036133.1 | 8 | 1129 | ||
| GenBank mRNA | AF071316.1 | - | 8 | 1131 | ||
| BC003724.1 | - | 7 | 1010 | 3593979 | ||
| CF898433.1 | - | 4 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cops7a| Gene Symbol: Cops7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012003.2 | NP_036133.1 | 8 | 1129 | ||
| GenBank mRNA | AF071316.1 | - | 8 | 1131 | ||
| BC003724.1 | - | 7 | 1010 | 3593979 | ||
| CF898433.1 | - | 4 | 625 | |||