siRNA Details
| Gene Symbol: Zfp346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012017.2 | NP_036147.1 | 7 | 1631 | ||
| GenBank mRNA | AF083339.1 | - | 7 | 1636 | ||
| AK028427.1 | - | 1 | 788 | |||
| AK084710.1 | - | 1 | 106 | |||
| BC055075.1 | - | 7 | 1625 | 30056643 | ||
| BU706820.1 | - | 1 | 286 | |||

Zfp346
Genomic Map