siRNA Details
| Gene Symbol: Trpc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF139923.1 | - | 9 | 2243 | ||
| AK135153.1 | - | 9 | 2432 | |||
| AK137418.1 | - | 10 | 2568 | |||
| BC117995.1 | - | 9 | 2243 | 40096022 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpc7| Gene Symbol: Trpc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF139923.1 | - | 9 | 2243 | ||
| AK135153.1 | - | 9 | 2432 | |||
| AK137418.1 | - | 10 | 2568 | |||
| BC117995.1 | - | 9 | 2243 | 40096022 | ||