siRNA Details
| Gene Symbol: Vat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012037.2 | NP_036167.1 | 5 | 1115 | ||
| GenBank mRNA | AK034795.1 | - | 5 | 1117 | ||
| AK145193.1 | - | 5 | 1149 | |||
| AK154178.1 | - | 1100 | ||||
| AK154965.1 | - | 5 | 1098 | |||
| AK155342.1 | - | 5 | 1117 | |||
| AK159310.1 | - | 5 | 1058 | |||
| AK159315.1 | - | 5 | 1100 | |||
| AK160891.1 | - | 5 | 1107 | |||
| AK170371.1 | - | 5 | 1096 | |||
| AK216339.1 | - | 1 | 104 | |||
| BC024586.1 | - | 3 | 344 | |||
| BC025496.1 | - | 5 | 1071 | 5320239 | ||
| BC036169.1 | - | 4 | 557 | |||
| BC085504.1 | - | 5 | 1034 | |||
| BC099678.1 | - | 5 | 1097 | 6311681 | ||

Vat1
Genomic Map