siRNA Details
| Gene Symbol: Vat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012037.2 | NP_036167.1 | 6 | 1869 | ||
| GenBank mRNA | AK034795.1 | - | 6 | 1871 | ||
| AK145193.1 | - | 6 | 1907 | |||
| AK154178.1 | - | 1954 | ||||
| AK154965.1 | - | 6 | 1853 | |||
| AK155342.1 | - | 6 | 1871 | |||
| AK159310.1 | - | 6 | 1813 | |||
| AK159315.1 | - | 6 | 1854 | |||
| AK160891.1 | - | 6 | 1861 | |||
| AK170371.1 | - | 6 | 1850 | |||
| AK195824.1 | - | 1 | 84 | |||
| AK200203.1 | - | 1 | 84 | |||
| AK210032.1 | - | 1 | 138 | |||
| BC024586.1 | - | 4 | 1100 | |||
| BC025496.1 | - | 6 | 1825 | 5320239 | ||
| BC036169.1 | - | 5 | 1311 | |||
| BC085504.1 | - | 6 | 1788 | |||
| BC099678.1 | - | 6 | 1851 | 6311681 | ||

Vat1
Genomic Map