siRNA Details
| Gene Symbol: ZPBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007009.2 | NP_008940.2 | 3, 4 | 397 | ||
| XM_011515095.2 | XP_011513397.1 | 3, 4 | 391 | |||
| XM_011515097.1 | XP_011513399.1 | 3, 4 | 391 | |||
| XM_011515102.2 | XP_011513404.1 | 3, 4 | 570 | |||
| XM_011515103.1 | XP_011513405.1 | 3, 4 | 391 | |||
| XM_017011707.1 | XP_016867196.1 | 3, 4 | 391 | |||
| GenBank mRNA | BC005223.1 | - | 3, 4 | 356 | 3950015 | |
| BT006822.1 | - | 3, 4 | 327 | |||
| D17570.1 | - | 3, 4 | 363 | |||

ZPBP
Genomic Map