siRNA Details
| Gene Symbol: Omd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012050.2 | NP_036180.1 | 3 | 1148 | ||
| GenBank mRNA | AB007848.1 | - | 3 | 1153 | ||
| AK050297.1 | - | 1 | 251 | |||
| BC095997.1 | - | 2 | 954 | 30739852 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Omd| Gene Symbol: Omd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012050.2 | NP_036180.1 | 3 | 1148 | ||
| GenBank mRNA | AB007848.1 | - | 3 | 1153 | ||
| AK050297.1 | - | 1 | 251 | |||
| BC095997.1 | - | 2 | 954 | 30739852 | ||