siRNA Details
| Gene Symbol: Tek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290549.1 | NP_001277478.1 | 2 | 682 | ||
| NM_001290551.1 | NP_001277480.1 | 2 | 682 | |||
| NM_013690.3 | NP_038718.2 | 2 | 682 | |||
| GenBank mRNA | BC050824.1 | - | 2 | 676 | 6333576 | |
| D13738.1 | - | 2 | 681 | |||
| S67051.1 | - | 3 | 345 | |||
| X67553.1 | - | 2 | 453 | |||
| X71426.1 | - | 3 | 416 | |||

Tek
Genomic Map