siRNA Details
| Gene Symbol: Tnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013694.4 | NP_038722.3 | 1 | 136 | ||
| GenBank mRNA | AK005689.1 | - | 1 | 141 | ||
| BC092529.1 | - | 1 | 136 | 6773112 | ||
| J03494.1 | - | 1 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnp2| Gene Symbol: Tnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013694.4 | NP_038722.3 | 1 | 136 | ||
| GenBank mRNA | AK005689.1 | - | 1 | 141 | ||
| BC092529.1 | - | 1 | 136 | 6773112 | ||
| J03494.1 | - | 1 | 111 | |||