siRNA Details
| Gene Symbol: Ubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083319.1 | NP_001076788.1 | 12, 13 | 1599 | ||
| NM_013699.2 | NP_038727.1 | 11, 12 | 1491 | |||
| GenBank mRNA | AB221581.1 | - | 10, 11 | 1079 | ||
| AK079815.1 | - | 4, 5 | 326 | |||
| AK140942.1 | - | 6, 7 | 496 | |||
| AK146040.1 | - | 1 | 111 | |||
| AY182062.1 | - | 11, 12 | 1188 | |||
| BC034749.1 | - | 10, 11 | 1818 | |||
| BC139828.1 | - | 10, 11 | 1108 | 40142981 | ||
| BC141082.1 | - | 10, 11 | 1170 | 9055694 | ||
| CD806234.1 | - | 6, 7 | 565 | |||
| CF172242.1 | - | 6, 7 | 593 | |||
| U20086.1 | - | 10, 11 | 1082 | |||

Ubp1
Genomic Map