siRNA Details
| Gene Symbol: GLIPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006851.2 | NP_006842.2 | 2 | 522 | ||
| XM_011537775.2 | XP_011536077.1 | 2 | 501 | |||
| XM_017018721.1 | XP_016874210.1 | 2 | 402 | |||
| GenBank mRNA | AF400440.1 | - | 2 | 402 | ||
| AK304847.1 | - | 2 | 502 | |||
| AK313449.1 | - | 2 | 388 | |||
| BC012510.1 | - | 2 | 438 | 4453841 | ||
| CB123261.1 | - | 2 | 575 | |||
| EF667354.1 | - | 2 | 374 | |||
| U16307.3 | - | 2 | 458 | |||
| X91911.1 | - | 2 | 502 | |||

GLIPR1
Genomic Map