siRNA Details
Gene Symbol: Fgd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159538.1 | NP_001153010.1 | 16 | 2255 | ||
NM_013710.4 | NP_038738.2 | 16 | 2252 | |||
GenBank mRNA | AF017368.1 | - | 16 | 2188 | ||
AK042260.1 | - | 16 | 2253 | |||
AK156151.1 | - | 16 | 2252 | |||
AK179355.1 | - | 1 | 147 | |||
BC021845.1 | - | 13 | 1711 | 4016697 |

More Information
Panther Classification:
Molecular Function -



Gene Ontology Categories:
