siRNA Details
| Gene Symbol: Itgb1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013712.2 | NP_038740.1 | 5, 6 | 481 | ||
| GenBank mRNA | AF140691.1 | - | 5, 6 | 425 | ||
| AK003906.1 | - | 5, 6 | 482 | |||
| BC030035.1 | - | 5, 6 | 456 | 1346742 | ||

Itgb1bp2
Genomic Map