siRNA Details
| Gene Symbol: Itgb1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013712.2 | NP_038740.1 | 8 | 679 | ||
| GenBank mRNA | AF140691.1 | - | 8 | 623 | ||
| AK003906.1 | - | 8 | 680 | |||
| BC030035.1 | - | 8 | 654 | 1346742 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgb1bp2| Gene Symbol: Itgb1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013712.2 | NP_038740.1 | 8 | 679 | ||
| GenBank mRNA | AF140691.1 | - | 8 | 623 | ||
| AK003906.1 | - | 8 | 680 | |||
| BC030035.1 | - | 8 | 654 | 1346742 | ||