siRNA Details
Gene Symbol: Krtap15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013713.1 | NP_038741.1 | 1 | 594 | ||
GenBank mRNA | AK132534.1 | - | 1 | 610 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013713.1 | NP_038741.1 | 1 | 594 | ||
GenBank mRNA | AK132534.1 | - | 1 | 610 |