siRNA Details
| Gene Symbol: Eif2ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177806.1 | NP_001171277.1 | 7 | 713 | ||
| NM_013719.3 | NP_038747.2 | 8 | 1017 | |||
| GenBank mRNA | AF193342.1 | - | 3 | 920 | ||
| AF193343.1 | - | 8 | 1009 | |||
| AF193344.1 | - | 8 | 1101 | |||
| AJ243533.1 | - | 8 | 1017 | |||
| AK129334.1 | - | 2 | 185 | |||
| BC023958.1 | - | 8 | 1040 | |||
| BC072637.1 | - | 7 | 713 | 30354433 | ||
| CO430666.1 | - | 4 | 372 | |||

Eif2ak4
Genomic Map