siRNA Details
| Gene Symbol: Chaf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013733.3 | NP_038761.1 | 3 | 277 | ||
| GenBank mRNA | AJ132771.2 | - | 3 | 181 | ||
| AK034839.1 | - | 3 | 278 | |||
| AK132551.1 | - | 101 | ||||
| AK135763.1 | - | 3 | 290 | |||
| BC053740.1 | - | 3 | 248 | 6504838 | ||

Chaf1a
Genomic Map