siRNA Details
| Gene Symbol: Chaf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013733.3 | NP_038761.1 | 3 | 884 | ||
| GenBank mRNA | AJ132771.2 | - | 3 | 788 | ||
| AK034839.1 | - | 3 | 885 | |||
| AK132551.1 | - | 708 | ||||
| AK135763.1 | - | 3 | 897 | |||
| BC053740.1 | - | 3 | 855 | 6504838 | ||

Chaf1a
Genomic Map