siRNA Details
| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013736.4 | NP_038764.2 | 11 | 3105 | ||
| GenBank mRNA | AK036592.1 | - | 11 | 3106 | ||
| AK042376.1 | - | 1 | 543 | |||
| AK147088.1 | - | 11 | 3099 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tceb3| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013736.4 | NP_038764.2 | 11 | 3105 | ||
| GenBank mRNA | AK036592.1 | - | 11 | 3106 | ||
| AK042376.1 | - | 1 | 543 | |||
| AK147088.1 | - | 11 | 3099 | |||