siRNA Details
| Gene Symbol: Cars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013742.5 | NP_038770.3 | 2 | 190 | ||
| GenBank mRNA | AJ276796.1 | - | 2 | 157 | ||
| AK033328.1 | - | 2 | 157 | |||
| AK135433.1 | - | 2 | 160 | |||
| BC054717.1 | - | 2 | 139 | 5686364 | ||
| BC058954.1 | - | 2 | 142 | 5707198 | ||
| BY021374.1 | - | 2 | 186 | |||
| BY253195.1 | - | 2 | 191 | |||

Cars
Genomic Map