siRNA Details
| Gene Symbol: Plekhb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163182.1 | NP_001156654.1 | 8 | 1073 | ||
| NM_001163183.1 | NP_001156655.1 | 8 | 1031 | |||
| NM_001163184.1 | NP_001156656.1 | 7 | 926 | |||
| NM_001163185.1 | NP_001156657.1 | 7 | 916 | |||
| NM_001163186.1 | NP_001156658.1 | 7 | 1022 | |||
| NM_001163187.1 | NP_001156659.1 | 6 | 917 | |||
| NM_001291336.1 | NP_001278265.1 | 6 | 886 | |||
| NM_013746.3 | NP_038774.1 | 9 | 1178 | |||
| GenBank mRNA | AF000272.1 | - | 9 | 1149 | ||
| AF071000.1 | - | 6 | 905 | |||
| AF100613.1 | - | 7 | 1010 | |||
| AF101053.1 | - | 8 | 1044 | |||
| AK005102.1 | - | 7 | 881 | |||
| AK141634.1 | - | 885 | ||||
| AK158172.1 | - | 7 | 917 | |||
| AK164683.1 | - | 7 | 881 | |||
| BC024756.1 | - | 9 | 1139 | 5369461 | ||

Plekhb1
Genomic Map