siRNA Details
| Gene Symbol: Phlda3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013750.2 | NP_038778.1 | 1 | 752 | ||
| GenBank mRNA | AK003767.1 | - | 1 | 718 | ||
| AK187696.1 | - | 1 | 61 | |||
| AK199905.1 | - | 1 | 52 | |||
| AK206046.1 | - | 1 | 46 | |||
| AK210974.1 | - | 1 | 46 | |||
| AK214255.1 | - | 1 | 46 | |||
| BC023376.1 | - | 1 | 654 | |||
| BC023408.1 | - | 1 | 654 | 5252055 | ||

Phlda3
Genomic Map