siRNA Details
| Gene Symbol: Pald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013753.2 | NP_038781.2 | 11 | 1556 | ||
| GenBank mRNA | AK088577.1 | - | 11 | 1392 | ||
| AK146489.1 | - | 11 | 1501 | |||
| BC137695.1 | - | 10 | 1320 | 8860714 | ||
| BC144846.1 | - | 10 | 1329 | 9053394 | ||
| BC144847.1 | - | 10 | 1326 | 9053395 | ||
| X99384.1 | - | 10 | 1341 | |||

Pald1
Genomic Map