siRNA Details
| Gene Symbol: Gyg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013755.3 | NP_038783.1 | 2 | 221 | ||
| GenBank mRNA | AA553085.1 | - | 2 | 220 | ||
| AF114031.1 | - | 2 | 156 | |||
| AK159576.1 | - | 2 | 202 | |||
| AK201125.1 | - | 1 | 129 | |||
| AK205608.1 | - | 2 | 119 | |||
| AK209105.1 | - | 2 | 138 | |||
| AK209409.1 | - | 2 | 119 | |||
| BC029903.1 | - | 2 | 182 | 4487769 | ||

Gyg
Genomic Map