siRNA Details
| Gene Symbol: Sh2d3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252547.1 | NP_001239476.1 | 11 | 2529 | ||
| NM_013781.3 | NP_038809.1 | 12 | 3015 | |||
| GenBank mRNA | AB030442.1 | - | 11 | 2492 | ||
| AB043953.1 | - | 12 | 2998 | |||
| AK042709.1 | - | 11 | 2513 | |||
| AK155165.1 | - | 12 | 2951 | |||
| AK172072.1 | - | 13 | 3394 | |||

Sh2d3c
Genomic Map