siRNA Details
| Gene Symbol: Ip6k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013785.2 | NP_038813.2 | 6 | 3163 | ||
| GenBank mRNA | AK004808.1 | - | 1 | 1391 | ||
| AK037925.1 | - | 5 | 2973 | |||
| AK088872.1 | - | 6 | 3181 | |||
| AK172921.1 | - | 6 | 3222 | |||
| BC053030.1 | - | 6 | 3167 | |||
| BC059006.1 | - | 6 | 3163 | 6827203 | ||

Ip6k1
Genomic Map