siRNA Details
| Gene Symbol: Tcf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114140.1 | NP_001107612.1 | 2 | 589 | ||
| NM_013836.3 | NP_038864.3 | 2 | 589 | |||
| GenBank mRNA | AY007594.1 | - | 2 | 589 | ||
| BC138038.1 | - | 2 | 562 | 8861057 | ||
| BC171946.1 | - | 2 | 562 | |||

Tcf20
Genomic Map