siRNA Details
| Gene Symbol: Zfp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044747.2 | NP_001038212.1 | 3 | 323 | ||
| NM_001163797.1 | NP_001157269.1 | 4 | 413 | |||
| NM_013844.2 | NP_038872.1 | 4 | 419 | |||
| GenBank mRNA | AB024005.1 | - | 4 | 403 | ||
| AK013868.1 | - | 4 | 414 | |||
| AK044831.1 | - | 3 | 293 | |||
| AK146092.1 | - | 4 | 380 | |||
| AK153887.1 | - | 4 | 355 | |||
| BC018171.1 | - | 4 | 402 | 3989444 | ||
| BY088951.1 | - | 3 | 326 | |||

Zfp68
Genomic Map